Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Luminaris Color HRM qPCR Master Mix

Luminaris Color HRM Master Mix is a convenient ready-to-use master mix designed for accurate detection of DNA sequence variations using High Resolution Melt analysis method.

$264.00 - $1026.00

Specifications

Concentration 2X
Detection Method HRM
Fidelity (vs. Taq) 2 X
GC-Rich PCR Performance High
PCR Method qPCR
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
FERK1031
View Documents
Thermo Scientific™
K1031BID
250 Reactions
Each for $264.00
Add to cart
 
FERK1032
View Documents
Thermo Scientific™
K1032BID
1250 Reactions
Each for $1,026.00
Add to cart
 
Description

Description

  • High performance on most real-time instruments with HRM capabilities
  • Dual-color system for easy reaction setup
  • Superior discrimination of all four SNP classes
  • High sensitivity - accurate results from 30 copies of genomic DNA
  • Exceptional stability of fully assembled reaction - for high-throughput applications
  • Optimized for PikoReal Real-Time System and software
Specifications

Specifications

2X
2 X
qPCR
HRM (High Resolution Melting) Analysis, Mutation Scanning
Luminaris
250 Reactions
HRM
High
Taq DNA Polymerase
SNPs (Unknown or Numerous), Epigenetic Targets
qPCR Master Mix
SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit