Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Luminaris Color HRM qPCR Master Mix

Luminaris Color HRM Master Mix is a convenient ready-to-use master mix designed for accurate detection of DNA sequence variations using High Resolution Melt analysis method.

Supplier:  Thermo Scientific™ K1031BID

 View more versions of this product

Catalog No. FERK1031

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Description

Description

  • High performance on most real-time instruments with HRM capabilities
  • Dual-color system for easy reaction setup
  • Superior discrimination of all four SNP classes
  • High sensitivity - accurate results from 30 copies of genomic DNA
  • Exceptional stability of fully assembled reaction - for high-throughput applications
  • Optimized for PikoReal Real-Time System and software
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 2 x 1.25 mL Luminaris Color HRM Master Mix (2X) containing chemically modified Hot Start Taq DNA Polymerase, EvaGreen™ fluorescent dye, dNTPs and MgCl2 in an optimized PCR buffer with a blue dye; sufficient for 250 x 20 μL reactions
• 1 x 1.25 mL 40X Yellow Sample Buffer
• 2 x 1.25 mL Nuclease-Free Water

Store at -20°C.
High
Taq DNA Polymerase
SNPs (Unknown or Numerous), Epigenetic Targets
qPCR Master Mix
2X
250 Reactions
HRM
qPCR
HRM (High Resolution Melting) Analysis, Mutation Scanning
Luminaris
250 Reactions
2 X
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.