Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Luminaris HiGreen qPCR Master Mix

Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.

Manufacturer:  Thermo Scientific™ K0991

 View more versions of this product

Catalog No. FERK0991


Add to cart

Includes: Luminaris HiGreen qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG, dNTPs (with dUTP) and SYBR Green I in an optimized PCR buffer; Nuclease-Free Water

Description

Description

Ready-to-use qPCR master mixes with premixed UDG optimized for SYBR Green chemistry using standard cycling protocol. Master mixes available with or without blue color.

  • UDG in the master mix to prevent carry-over contamination
  • Equal performance of blue and colorless master mixes
  • Versions with premixed ROX or fluorescein for different real-time platforms
  • Standard cycling protocols
  • Excellent qPCR performance
  • Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity: single copy detection
  • Wide linear range: accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience: ready-to-use 2X master mix
Specifications

Specifications

250 x 20μL rxns
Store at -20°C.
Detection Method: SYBR, PCR Method: qPCR
Bio-Rad CFX384 Touch™, BioRad CFX96™, Cepheid SmartCycler, Corbett Rotor-Gene, Eppendorf MasterCycler RealPlex, MJ Chromo4, MJ Opticon, Roche LightCycler 480
250 Reactions
DNA, dsDNA
2X
Real time PCR (qPCR), gene expression analysis, siRNA validation, genotyping and pathogen detection
Luminaris HiGreen qPCR Master Mix (2X) includes Hot Start Taq DNA Polymerase, UDG and dNTPs (also dUTP) in an optimized PCR buffer; supplied with nuclease-free water.
Safety and Handling

Safety and Handling

ShelfLife : 2 Years

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.