Learn More
Thermo Scientific™ MagJET™ RNA Kit
Uses magnetic bead technology to purify total RNA from cultured mammalian cells, tissues, bacteria, and yeast.
Manufacturer:  Thermo Scientific™ K2732
 View more versions of this product
Catalog No. FERK2732
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The Thermo Scientific MagJET RNA Kit is designed for fast and efficient purification of total RNA from cultured mammalian cells, tissues, bacteria and yeast.
The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual nucleic acid purification by low sample throughput users.
The resulting high quality purified RNA is free of proteins, nucleases and other contaminants or inhibitors, can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR and other enzymatic reactions. See Table 1 for typical total RNA yields from various sources.
Highlights
·Versatile - suitable for total RNA purification from variety of biological samples, including eukaryotic cells, animal tissues, yeast and bacteria
·Efficient - high yields of high-integrity RNA
·High quality - isolates intact total RNA, free of inhibitors and gDNA contamination
·Flexible - easily adjustable for high throughput automation platforms and manual processing
Applications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
· RT-PCR
· RT-qPCR
· Northern blotting
· Nuclease protection assay
Includes
· Lysis Buffer
· MagJET Magnetic Beads
· DNase I (lyophilized)
· 10X Reaction Buffer with MgCl2 for DNase I
· DNase I Reconstitution Buffer(conc.)
· Wash Buffer 1 (conc.)
· Wash Buffer 2 (conc.)
· Water, nuclease free
Hazardous:Yes
> Quality Control:The MagJET RNA Kit is qualified by isolating RNA from 5 mg of frozen mouse muscle or heart following the protocol outlined in the manual. The quality of isolated RNA is evaluated spectrophotometrically and by agarose gel electrophoresis. The purified RNA has an A260/A280 ratio of 2.0±0.2. The functional quality of purified RNA is evaluated by RT-PCR.
> Shelf Life:24 Months
> Shipping Condition:Ambient
> Storage Condition:Ambient
Specifications
Specifications
High purity RNA suitable for all conventional molecular biology procedures like RT-PCR, RT-qPCR, Northern blotting, Nuclease protection assay
Lysis buffer (200mL), MagJET magnetic beads (2 x 8.5mL), lyophilized DNase I (1 vial), 10x reaction buffer with MgCl2 for DNase I (9 x 1mL), Conc. DNase I reconstitution buffer (2.5mL), conc. wash buffer 1 (2 x 80mL), conc. wash buffer 2 (3 x 45mL), nuclease free water (125mL)
4 x 96 Preps
Store lyophilized DNase I, 10x Reaction Buffer with MgCl2 for DNase I and DNase I Reconstitution Buffer at -20deg.C upon arrival. Store MagJET Magnetic Beads at 4deg.C and other components of the kit at room temperature (15deg. to 25deg.C)
Safety and Handling
Safety and Handling
ShelfLife : 2 Years