Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TopVision Agarose Tablets

High quality, multipurpose agarose tablets in blister packaging.

Manufacturer:  Thermo Scientific™ R2801

 View more versions of this product

Catalog No. FERR2801


Add to cart

Description

Description

Description
TopVision Agarose Tablets, low EEO (LE), is multipurpose agarose in tablet format delivered in a convenient blister pack. The agarose tablets are made of standard melting point agarose that yields high resolution, sharp DNA bands with high clarity and low background. Its optimized gel strength enhances ease of gel processing and handling. Manufactured using an innovative Organic Solvent-Free manufacturing process that is greener and more environmentally friendly.
Features
· Convenient tablet format - no messy weighing required.
· Fast dissolving protocol.
·“Green”- manufactured with significantly reduced environmental impact, using an innovative organic solvent-free technology.
· Enhanced resolution and reduced background.
Applications
Ideal for routine DNA and RNA gel electrophoresis and blotting assays.
Environmentally responsible Green Solution
Unlike traditional agarose powder, which production relies heavily on the use of toxic organic solvents, TopVision Agarose Tablets are manufactured using an innovative process that completely eliminates dependency on organic solvents and significantly reduces environmental impact while creating a safer working environment.

Appearance:White to off-white powder
> Ash:≤ 0.5%
> CAS:9012-36-6
> EEO:≤ 0.13
> Gel Strength:≥ 1200 g/cm2 (1% Gel)
> Gelling Point:36°C ± 1.5°C (1.5% gel)
> Hazardous:No
> Impurities (DNAses, RNases and Proteases):None
> Melting Point:88°C ± 1.5°C (1.5% gel)
> Moisture:≤ 10%
> Shelf Life:24 Months
> Shipping Condition:Ambient
> Solubility:Clear colorless solution at 1g in 100 mL water
> Storage Condition:Ambient
> Sulfate:≤ 0.15%

Specifications

Specifications

For routine DNA and RNA gel electrophoresis and blotting assays
200 Tablets (0.5g Each)
Ambient
Safety and Handling

Safety and Handling

ShelfLife : 2 years

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit