Learn More
Thermo Scientific™ Finntip™ Filtered Pipet Tips, Nonsterile
Effectively eliminate carryover contamination from liquids and aerosol
Manufacturer:  Thermo Scientific™ 94052210
 View more versions of this product
Catalog No. 14-386-200
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
All racks are color coded to ensure precise matching with the right size Thermo Scientific Finnpipette pipettors.
  • Combines the ergonomics of a standard Finntip Flex pipette tip with the safety of a filter tip
  • Free from DNA, DNase, and RNase - ideal for use in PCR, research applications, and pipetting radioactive or infections samples
  • Flex tips load and eject easily for reduced hand fatigue
Specifications
Specifications
100μL
Filtered
Non-sterile