Learn More
Thermo Scientific™ Finntip™ Pocket Tips Available on GSA/VA Contract for Federal Government customers only.
Dispense nanoliter volumes with handheld pipettes
Manufacturer:  Thermo Scientific™ 94035550
 View more versions of this product
Catalog No. 14-487-87
Add to cart
Description
Description
Tips have a molded pocket inside to allow transfer of compounds suspended in DMSO into an assay.
  • 50 and 250nl dispensing
  • Works with DMSO solution
  • Reduces reagent costs
  • Eliminates the need for intermediate dilutions
  • Provides excellent mixing
  • Length: 53mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Rack
250nL