Learn More
Thermo Scientific™ FirstDefender™ RM Handheld Chemical Identification
Enhanced speed, performance, mixture analysis, and user interface make this Raman ideal for rapid, accurate identification of unknown chemicals directly in the field
Supplier:  Thermo Scientific™ 8000017401
 View more versions of this product
Catalog No. 17-720-412
Add to Cart
Description
Description
  • Fast, accurate identification based on Raman spectroscopy to quickly identify unknown solid and liquid chemicals down range
  • Sophisticated chemometric algorithms automatically determine presence of mixed chemicals
  • The easy-to-navigate and extensive substance library includes explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more
  • Lightweight, ergonomic design weighs 1.8 lb. (800g) for easy transportation down range
  • Point-and-shoot™ identification operates directly through sealed translucent glass or plastic containers, avoiding exposure to potentially harmful substances; an integrated vial holder offers an alternative sampling method
  • Easy to use and maintain with a menu driven user interface that requires minimal training and no scheduled maintenance or calibration
  • Nondestructive testing—optical technique maintains sample integrity
  • Can be powered using the included rechargeable battery, wall adapter, or SureFire™ disposable batteries
Certifications
Certified to MIL-STD 810G standards for ruggedness and environmental protection to IP67 standards against dust and water up to 1m.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4-year warranty and support
Product Suggestions
Product Suggestions