Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Flexaframe™ Multiclutch Connectors
Permit lattice adjustment after construction
Description
- Hold Flexaframe rods at 90° angle in lattice
- Allow horizontal and vertical adjustment after lattice is built
- Two adjacent rods fit into oval hole in connector, prevent turning when tightened
- 0.5 in. diameter (13mm) holes in side and front of connector allow many different rod configurations
- Castaloy™ metal
Specifications
Metal | |
Multiclutch Connector | |
Permits lattice adjustment after construction, holds rods at 90° angle in lattice |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction