Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer COD Vial Cell Holder
Manufacturer:  Thermo Scientific™ 4014000
Catalog No. 14-385-449
Add to cart
Description
Description
COD Vial Cell Holder. Snap-in” holder can handle one COD vial up to 104mm tall and 18mm in diameter.
Specifications
Specifications
Genesys™ 20 spectrophotometer
One COD vial up to 104mm tall and 18mm dia.
104mm
Cell Holder