Thermo Scientific™ Gill Hematoxylins

Rapid-staining, usually requiring an immersion time of two minutes or less

Manufacturer: Thermo Scientific™ 1931403

 View more versions of this product

Catalog No. 14-390-11

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


    Specifications

    For Use With (Application) For delicate cytological staining
    Shelf Life 2 Years
    Type Gill 1 Hematoxylin
    Quantity 1 gal.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit