Learn More
Thermo Scientific™ Gill Hematoxylins
Rapid-staining, usually requiring an immersion time of two minutes or less
Manufacturer:  Thermo Scientific™ 1931432
 View more versions of this product
Catalog No. 14-390-16
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications Description & Specifications
Specifications
Gill 3 Hematoxylin
4 x 16 oz.
For delicate cytological staining
Safety and Handling
ShelfLife : 2 Years
Accessories