Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Golden Gate ATR for Nicolet™ iS5 FT-IR Spectrometer

This versatile infrared sampling accessory is fast, sensitive, and robust

Manufacturer:  Thermo Scientific™ 869145200

 View more versions of this product

Catalog No. 14-387-612


Add to cart

Requires : Nicolet iS5 FT-IR Spectrometer (Sold Separately)

Description

Description

Analyzes sample types from hard solids to corrosive liquids

  • Used to analyze a range of samples from single particles and fibers to corrosive liquids
  • High temperature bonded into its tungsten carbide mount giving performance and strength to withstand the high pressures
  • Used for almost any sample type in any research or QA environment
  • Little or no sample preparation needed
  • Valuable accessory for the preparation of toxic compounds
  • Ideal for macro sampling
  • 45° single reflection Type IIa diamond
  • Brazed tungsten carbide diamond mount
  • Fully enclosed beam condensing lens system
  • Sapphire anvil for high pressure contact
  • Reproducible pressure control to 80 pounds
  • Thermal, cryogenic, and super critical fluid options available
  • Fully enclosed sample anvil available for dangerous or sensitive samples
Specifications
  • Crystal material: Type IIa diamond
  • Spectral wavelength range: 5000 to 650cm–1 ZnSe optics, 5000 to 400cm–1 KRS–5 optics

Polymer pellets, intractable or opaque solids, single fibers, corrosive liquids, geochemical samples, powders, coated wires, air sensitive samples, hazardous materials/toxic compounds

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit