Learn More
Thermo Scientific H-6000A Bucket Adapters
Accept a variety of blood processing tubes, bags and bottles
Manufacturer: Thermo Scientific 01092
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
- Adapters hold a maximum capacity of 6 x 500mL (bottles), 6 x 550mL (blood bags), and 132 x 7mL (tubes). Max. O.D. x L: 70 x 160mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.