Learn More
Thermo Scientific™ H-6000A Bucket Adapters Available on GSA/VA Contract for Federal Government customers only.
Accept a variety of blood processing tubes, bags and bottles
Manufacturer:  Thermo Scientific™ 00436
 View more versions of this product
Catalog No. 00-436
Add to cart
Description
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
  • Adapters hold a maximum capacity of 6 x 500mL (bottles), 6 x 550mL (blood bags), and 132 x 7mL (tubes). Max. O.D. x L: 70 x 160mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor