Thermo Scientific™ HAAKE™ Viscotester™ E, D and C

Ready to go package—unpack, switch on and start measuring viscosity

Manufacturer: Thermo Scientific™ 3990400

 View more versions of this product

Catalog No. 13-874-594

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance BS: 6075, 5350, ISO: 2555, 1652, ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716
    Hertz 50/60Hz
    Voltage 100/240V
    Speed Range 0.1 to 200rpm
    Count (Speed Setting) 21
    Display Type LCD
    Electrical Requirements 100/240V 50/60Hz
    For Use With (Application) Routine viscosity measurements with data transfer
    Includes Spindle R2 to R7
    Precision ±1%
    Probe Type PT100
    Product Type Viscotester D Rotational Viscometer
    Repeatability 0.2%
    Viscosity 100 to 13.000.000mPas

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit