Learn More
Thermo Scientific™ HAAKE™ Viscotester™ E, D and C
Ready to go package—unpack, switch on and start measuring viscosity
Manufacturer:  Thermo Scientific™ 3990501
 View more versions of this product
Catalog No. 13-874-595
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
54
100/240V 50/60Hz
Spindle L1 to L4
Viscotester E Rotational Viscometer
15 to 6.000.000mPas
100/240V
BS: 6075, 5350, ISO: 2555, 1652, ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716
LCD
Automatic flow curve measurements with full PC control
±1%
0.2%
50/60Hz
0.01 to 200rpm