Thermo Scientific™ Halt™ Phosphatase Inhibitor Cocktail

Safeguards against serine, threonine and protein tyrosine phosphatases activity

The Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases as well as protein tyrosine phosphatases (PTPs). The cocktail contains a mixture of four inhibitors of broad specificity, including sodium fluoride, sodium orthovanadate, sodium pyrophosphate, and β-glycerophosphate. Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract. The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays. Furthermore, the cocktail is compatible with popular protein assays such as the Thermo Scientific Pierce™ BCA Protein Assay Kit (Part No. 23225) and Bradford-based Pierce Coomassie Plus™ Assay Kit (Part No. 23236).

$192.19 - $789.14

Specifications

Format Liquid
Formulation Inhibitors stabilized in aqueous solution
View More Specs
  • Products 4
  • Description & Specifications
Products
Catalog Number Mfr. No. Sufficient For Quantity Price Quantity & Availability    

PI-78420

Thermo Scientific
78420
100mL of sample 1mL Each for $192.19
Check Availability
Add to Cart

PI-78426

Thermo Scientific
78426
500mL of sample 5 x 1mL Each for $585.81
Check Availability
Add to Cart

PI-78427

Thermo Scientific
78427
1L of sample 10mL Each for $789.14
Check Availability
Add to Cart

PI-78428

Thermo Scientific
78428
240mL of sample in 10mL increments 24 x 100µL Each for $298.25
Check Availability
Add to Cart
Descriptions & Specifications

Related Products