Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Heat-Shrink Bands for Nalgene™ PETG Media Bottles
Tamper-resistant seal
$153.05 - $269.45
Specifications
Quantity 1000
View More Specs
  • Products 4
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
01-814-50
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
312160-0200
Heat-shrink band for 30mL PETG Square Media Bottle with 24-415 HDPE Closure
Case of 1000 for $153.05
Check Availability
Add to Cart
01-814-51
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
312160-0240
Heat-shrink band for 60mL PETG Square Media Bottle with 24-415 HDPE Closure
Case of 1000 for $181.56
Check Availability
Add to Cart
01-814-52
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
312160-0384
Heat-shrink band for 125-1000mL PETG Square Media Bottle with 38-430 HDPE Closure
Case of 1000 for $212.53
Check Availability
Add to Cart
01-814-53
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
312160-0530
Heat-shrink band for 2000mL PETG Square Media Bottle with 53B HDPE Closure
Case of 1000 for $269.45
Check Availability
Add to Cart
Descriptions & Specifications
Related Products