Thermo Scientific™ Heracell™ 150i Tri-Gas Incubator Packages
Choice of noncorrosive stainless steel or 100% pure copper
Manufacturer: Thermo Scientific™ 50115058PR
 View more versions of this product
Catalog No. 11-687-201PR
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Chamber Material Copper
    Voltage 120V
    Controller Type Touchscreen
    Sensor Types TC sensor
    Warranty 2 Years
    Thermo Scientific Heracell i tri-gas incubators feature innovative ContraCon moist heat decontamination technology that ensures worry-free cleaning and operation. These incubators are designed to provide optimal growth conditions and deliver superior parameter recovery rates.
    iCAN™ Touchscreen Control
    • Door-mounted for easy accessibility and viewing
    • Convenient on-screen user prompts for simplified measurement and control navigation
    • Choice of languages
    • Changes in culture environment visible on screen logs and usage recording
    • Visual monitor alarm
    TC or IR CO2 Sensor
    • Standard thermal conductivity (TC) sensor features accurate gas sensor technology
    • Infrared (IR) sensor features exclusive split-beam technology for precise CO2 control in fluctuating temperature and humidity conditions
    Rapid Humidity Recovery
    • Large, directly heated water reservoir
    • Recovery times are up to five times faster than conventional water pans
    • Water level alarm with audible and visual alerts
    Optional O2 Control with Auto-Cal
    • Two O2 control ranges precisely control oxygen concentration
    • Ideal for applications requiring decreased or increased O2 conditions
    • Zirconium Oxide sensor
  • Description & Specifications
Related Products
Product Content Feedback