Learn More
Thermo Scientific™ Heracell™l VIOS 160i CO2 Incubator Packages Available on GSA/VA Contract for Federal Government customers only.
Achieve your next breakthrough
Manufacturer:  Thermo Scientific™ 51030420
 View more versions of this product
Catalog No. 13-998-141
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
100% copper
3°C above ambient to 55°C
905mm
637mm
790mm
504mm
465mm
423mm
216 lb.
18.3 in.
120V
USB, and optional 4-20mA
10/30kg
42mm access port, rear left
165L
120V 50/60Hz
183 lb.
Heracell VIOS 160i CO2 incubator single chamber with 4-20 mAMP data output
Integrated reservoir
35.6 in.
25.1 in.
31.1 in.
19.9 in.
18.3 in.
16.7 in.
83kg
98kg
465mm
4.6A during incubation, 8.3A during sterilization
22/66 lb.
50/60Hz
5.8. cu. ft.
CE, CSA
IR sensor
>93% @ 37°C