Learn More
Thermo Scientific™ Heracell™l VIOS 160i CO2 Incubator Packages Available on GSA/VA Contract for Federal Government customers only.
Achieve your next breakthrough
Manufacturer:  Thermo Scientific™ 51030414
 View more versions of this product
Catalog No. 13-998-136
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Stainless steel
3°C above ambient to 55°C
905mm
637mm
790mm
504mm
465mm
423mm
216 lb.
18.3 in.
120V
USB, and optional 4-20mA
10/30kg
42mm access port, rear left
165L
120V 50/60Hz
183 lb.
Heracell VIOS 160i CO2 incubator single chamber with internal CO2 gas guard
Integrated reservoir
35.6 in.
25.1 in.
31.1 in.
19.9 in.
18.3 in.
16.7 in.
83kg
98kg
465mm
4.6A during incubation, 8.3A during sterilization
22/66 lb.
50/60Hz
5.8. cu. ft.
CE, CSA
IR sensor
>93% @ 37°C