Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ High Purity Single Stage Brass Gas Regulators
Recommended for applications where precise flow control is not critical
Includes: Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Products 9
Catalog Number Mfr. No. Range Type Price Quantity & Availability  
Catalog Number Mfr. No. Range Type Price Quantity & Availability  
60-180-901
View Documents
Thermo Scientific™
60180901
0 to 200 psig CGA 580 with Triple Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
60-180-903
View Documents
Thermo Scientific™
60180903
0 to 200 psig DIN 6 with Triple Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
60-180-905
View Documents
Thermo Scientific™
60180905
0 to 200 psig CGA 350 with Triple Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
60-180-909
View Documents
Thermo Scientific™
60180909
0 to 200 psig CGA 320 with Triple Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
60-180-915
View Documents
Thermo Scientific™
60180915
0 to 200 psig CGA 580 with Combi Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
60-180-917
View Documents
Thermo Scientific™
60180917
0 to 200 psig DIN 6 with Combi Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
60-180-919
View Documents
Thermo Scientific™
60180919
0 to 200 psig CGA 350 with Combi Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
60-180-921
View Documents
Thermo Scientific™
60180921
0 to 200 psig DIN 1 with Combi Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
60-180-923
View Documents
Thermo Scientific™
60180923
0 to 200 psig CGA 320 with Combi Filter This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • For non-corrosive analytical and process applications
  • Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
  • Specially designed, convoluted, stainless steel diaphragm provides good regulating performance and maximum purity integrity without the need for a soft seal
  • Capable of withstanding an internal vacuum
  • All regulators supplied with 1/8 in. stainless steel output adaptor
  • Built in check valve allows for cylinder change without contaminating the gas line