Thermo Scientific™ High Purity Single Stage Brass Gas Regulators Available on GSA/VA Contract for Federal Government customers only.
Recommended for applications where precise flow control is not critical
Manufacturer: Thermo Scientific™ 60180-907
 View all options for this product
Catalog No. 60-180-907
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    • For non-corrosive analytical and process applications
    • Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
    • Specially designed, convoluted, stainless steel diaphragm provides good regulating performance and maximum purity integrity without the need for a soft seal
    • Capable of withstanding an internal vacuum
    • All regulators supplied with 1/8 in. stainless steel output adaptor
    • Built in check valve allows for cylinder change without contaminating the gas line
  • Description & Specifications
Related Products
Product Content Feedback