Learn More
Thermo Scientific™ High Purity Two Stage Brass Gas Regulators
Recommended for non-corrosive analytical and process applications requiring precise, stable, delivery pressure control
$1863.00
Specifications
Range | 0 to 200 psig |
---|
Includes: Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Catalog Number | Mfr. No. | Range | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Range | Type | Price | Quantity & Availability | ||||
60-180-926
|
Thermo Scientific™
60180926 |
0 to 200 psig | CGA 590 with Combi Filter |
Each for $1,863.00
|
|
||||
60-180-902
|
Thermo Scientific™
60180902 |
0 to 200 psig | CGA 580 with Triple Filter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
60-180-908
|
Thermo Scientific™
60180908 |
0 to 200 psig | DIN 1 with Triple Filter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
60-180-906
|
Thermo Scientific™
60180906 |
0 to 200 psig | CGA 350 with Triple Filter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
60-180-910
|
Thermo Scientific™
60180910 |
0 to 200 psig | CGA 320 with Triple Filter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
60-180-918
|
Thermo Scientific™
60180918 |
0 to 200 psig | DIN 6 with Combi Filter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
60-180-922
|
Thermo Scientific™
60180922 |
0 to 200 psig | DIN 1 with Combi Filter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
60-180-920
|
Thermo Scientific™
60180920 |
0 to 200 psig | CGA 350 with Combi Filter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
60-180-924
|
Thermo Scientific™
60180924 |
0 to 200 psig | CGA 320 with Combi Filter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- For non-corrosive analytical and process applications
- Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
- Specially designed, convoluted, stainless steel diaphragm provides goodregulating performance and maximum purity integrity without the needfor a soft seal
- Capable of withstanding an internal vacuum
- All regulators supplied with 1/8 in. stainless steel output adaptor
- Built in check valve allows for cylinder change without contaminating the gas line
Specifications
0 to 200 psig |