Learn More
Thermo Scientific™ High Purity Two Stage Brass Gas Regulators
Recommended for non-corrosive analytical and process applications requiring precise, stable, delivery pressure control
Supplier:  Thermo Scientific™ 60180902
 View more versions of this product
Catalog No. 60-180-902
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes: Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Description
Description
  • For non-corrosive analytical and process applications
  • Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
  • Specially designed, convoluted, stainless steel diaphragm provides goodregulating performance and maximum purity integrity without the needfor a soft seal
  • Capable of withstanding an internal vacuum
  • All regulators supplied with 1/8 in. stainless steel output adaptor
  • Built in check valve allows for cylinder change without contaminating the gas line
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0 to 200 psig
CGA 580 with Triple Filter
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories