Learn More
Thermo Scientific HistoScreen Cassettes in Tube Packs
Handle small biopsies and fragile specimens with Thermo Scientific HistoScreen Cassettes in Tube Packs.
Manufacturer: Thermo Scientific C750GY
Description
Handle small biopsies and fragile specimens with Thermo Scientific HistoScreen Cassettes in Tube Packs. Designed for automatic cassette labelers, they are the ultimate tube-fed cassettes for specimen security.
- Saves time, offers the greatest specimen security, and provides ease of use
- Provides an inner chamber lined with a “see-through” mesh for easy identification of specimens
- Eliminates the need for messy sponges or lens paper
- Ensures your tissues are safely processed with cassettes which will not pop open during processing
Specifications
HistoScreen™ Cassettes, Mesh Pore Insert | |
Gray |