Learn More
Thermo Scientific Hypercarb Javelin Guard HPLC Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
$871.00
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
35005024006
|
Thermo Scientific
35005024006 |
4 mm | 20 mm |
Pack of 2 for $871.00
|
|
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- Suited to solve problem separations
- Exceptional retention of very polar analytes
- True orthoganality to C18
- Separate structurally related substances
- Excellent for high temperature applications
Specifications
5 µm | |
Hypercarb |