Learn More
Thermo Scientific™ Hypersil™ BDS C18 Javelin Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Manufacturer:  Thermo Scientific™ 28105-011006
 View more versions of this product
Catalog No. 28105011006
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
5μm
10mm
C18
1mm