Thermo Scientific™ Hypersil™ BDS C18 Javelin Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Manufacturer: Thermo Scientific™ 28105-013006
 View all options for this product
Catalog No. 28105013006
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Particle Size 5μm
    Length (Metric) 10mm
    Stationary Phase C18
    Diameter (Metric) Inner 3mm
    • Direct-connection, fingertight design for convenience and maximum efficiency
    • Patented design requires no holder, allowing quick and easy replacement
    Features of the BDS C18 Solid Phase
    • Base deactivated with minimal residual silanol activity
    • Economical with exceptional reproducibility
  • Description & Specifications
Related Products
Product Content Feedback