Learn More
Thermo Scientific™ Hypersil™ BDS C18 Javelin Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Manufacturer:  Thermo Scientific™ 28103023006
 View more versions of this product
Catalog No. 28103023006
Add to cart
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of the BDS C18 Solid Phase
  • Base deactivated with minimal residual silanol activity
  • Economical with exceptional reproducibility
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
Specifications
Specifications
3μm
20mm
C18
3mm