Learn More
Thermo Scientific™ Hypersil™ BDS C8 Columns
High quality base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention
Manufacturer:  Thermo Scientific™ 28203053030
Catalog No. 28203053030
Add to cart
Description
Description
  • Excellent choice for acids, bases and neutral compounds
  • Less retentive than C18 for faster analyses
  • Robust columns with long column lifetimes
  • Very reliable for routine assays
Order Info
Other column dimensions are available. Please call Customer Service for more information.
Specifications
Specifications
3 mm
50 mm
3 µm
C8