Learn More
Thermo Scientific™ Hypersil™ BDS Phenyl Columns
Exceptional stability and alternative selectivity to C18 and C8 columns
$714.50
Specifications
Particle Size 5 μm
Stationary Phase Phenyl (Ph)
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity & Availability  
03-150-506
View Documents
Thermo Scientific™
28905102130
2.1 mm 100 mm
Each for $714.50
Add to cart
Description
Description
  • High quality, base-deactivated columns
  • Unique selectivity for aromatic and slightly polar compounds
  • Manufactured under ISO 9001:2000 conditions
Order Info
Other column dimensions are available. Please call your Customer Service Representative for more information.
Compliance
Manufactured under ISO 9001:2000 conditions.
Specifications
Specifications
5 μm
Phenyl (Ph)