Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypersil™ BDS Phenyl (Ph) Reversed Phase HPLC Column, 5 μm, 2.1 mm x 20 mm

Column Format: DASH-HTS Column, 1/pk.; Carbon Load: 5%; Pore Size: 130 Å; Column Phase: Reversed Phase

Supplier:  Thermo Scientific™ 28905022150

Catalog No. 28905022150

Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.


Description

Description

  • Patented, low dead volume design minimizes band spreading
  • Finger-tight fittings make changing columns quick and easy
  • Available in packs of 3 or packs of 10 for better cost-effectiveness

Features of BDS Phenyl Solid Phase

  • May be used to accomplish difficult separations
  • High quality, base deactivated
  • Selectivity for aromatic and slightly polar compounds
  • Manufactured under ISO 9001:2000 conditions

Specifications

Specifications

DASH-HTS Column, 1/Pk.
20 mm
1 to 9
L11
Base-Deactivated Silica
Reversed Phase
2.1 mm
5 μm
130 Å
Phenyl (Ph)
5%
Reversed Phase
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit