Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Hypersil C4 CPG HPLC Columns--3μm Particle Size
Supplier: Thermo Scientific™ 30403032131
Description
Specifications
2.1 mm | |
3 μm | |
C4 | |
Reversed Phase |
30 mm | |
L26 | |
Silica | |
Reversed Phase |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction