Learn More
Thermo Scientific™ Hypersil™ GOLD C8 KAPPA Capillary Columns
Narrow ID columns operate at low flow rates required for greater ionization efficiencies
Manufacturer:  Thermo Scientific™ 25203100565
 View more versions of this product
Catalog No. 25-203-100565
Add to cart
Description
Description
Features of Hypersil GOLD C8 Solid Phase
  • Hypersil GOLD C8 columns provide similar selectivity to C18 columns but with reduced retention
  • Lower hydrophobicity
  • Faster separations
  • Excellent peak shape
  • High efficiency
  • Outstanding sensitivity
Specifications
Specifications
0.5 mm
100 mm
3 µm
C8