Learn More
Thermo Scientific™ Hypersil™ GOLD CN Preparative HPLC Columns, 5µm Particle Size
Preparative columns are available in popular Thermo Scientific phases
Manufacturer:  Thermo Scientific™ 25805259370
 View more versions of this product
Catalog No. 03-051-150
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Hypersil GOLD CN columns provide alternative selectivity with lower hydrophobicity
  • Excellent peak shape
  • Outstanding senstivity
  • Less retention for faster analysis
Order Info
Other custom column dimensions are available. Please call your local Customer Service for more information.
Specifications
Specifications
30 mm
250 mm
5 µm
CN