Learn More
Thermo Scientific™ Hypersil™ ODS (C18) Columns, 10µm Particle Size Available on GSA/VA Contract for Federal Government customers only.
Provide an excellent C18 phase for a broad range of applications and a global standard for many existing methods
Manufacturer:  Thermo Scientific™ 30110259070
 View more versions of this product
Catalog No. 30110259070
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10mm
250mm
10μm
C18