Learn More
Thermo Scientific™ Hypersil™ SAS C1 Columns, 5µm Particle Size
The least retentive of the Hypersil alkyl bonded phases
Manufacturer:  Thermo Scientific™ 30505102130
 View more versions of this product
Catalog No. 03-150-575
Part Not Found
Description
Description
Order Info
Other column dimensions are available. Also available in 10µm. Please call Customer Service for more information.
Specifications
Specifications
2.1 mm
100 mm
5 µm
C1