Learn More
Thermo Scientific™ Hypersil™ SAX Columns, 5µm Particle Size Available on GSA/VA Contract for Federal Government customers only.
Highly stable silica-based quarternary amine strong anion exchange columns, designed for aqueous and low pH mobile phases
Manufacturer:  Thermo Scientific™ 34105154030
 View more versions of this product
Catalog No. 03-170-312
  Check Availability
Add to cart
Description
Description
  • High stability to aqueous and low pH mobile phases
  • Suited to the analysis of smaller organic molecules including nucleotides and organic acids
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Other column dimensions are available. Also available in 10µm. Please call your Customer Service Representative for more information.
Specifications
Specifications
4mm
150mm
5μm
SAX