Learn More
Thermo Scientific™ Hypersil™ Silica Javelin™ Guard Columns
Javelin guard columns offer convenient, easy-to-use protection for analytical HPLC columns
Manufacturer:  Thermo Scientific™ 30005024006
Catalog No. 30005024006
Add to cart
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Silica Solid Phase
  • Excellent batch-to-batch reproducibility
  • Spherical particle with narrow particle size distribution
  • Long column lifetime
  • High performance and column efficiency
Specifications
Specifications
5μm
20mm
4mm