Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyPURITY™ C4 Reversed Phase HPLC Column, 5 µm, 2.1 mm x 10 mm

Column Format: DASH-HTS Column; Carbon Load: 4.5%; Pore Size: 190 Å; Column Phase: Reversed Phase

Manufacturer:  Thermo Scientific™ 22405022150

Catalog No. 22405022150

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Patented, low dead volume design minimizes band spreading
  • Finger-tight fittings make changing columns quick and easy
  • Available in packs of 3 or packs of 10 for better cost-effectiveness

Specifications

Specifications

Silica
Reversed Phase
Reversed Phase
2.1 mm
5 μm
200 m²/g
C4
4.5%
Yes
DASH-HTS Column
10 mm
190 Å
L26
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit