Learn More
Thermo Scientific HyPURITY C4 Guard Cartridge
Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime
Manufacturer: Thermo Scientific 22405013001
Requires : Thermo Scientific UNIGUARD direct connection or stand alone holder, sold separately.
Description
Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime.
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD direct connection and stand alone holders
- High quality, highly pure silica columns
Specifications
3 mm | |
10 mm | |
5 µm | |
C4 |