Learn More
Thermo Scientific™ HyPURITY™ C8 Guard Cartridges
Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime
Manufacturer:  Thermo Scientific™ 22205012101
Catalog No. 22205012101
Add to cart
Description
Description
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • High quality, highly pure silica columns
Specifications
Specifications
5μm
10mm
2.1mm
C8
Documents
Accessories