Thermo Scientific™ HyPURITY™ Cyano Guard Cartridge Available on GSA/VA Contract for Federal Government customers only.

Protect the high quality, highly pure silica Hypurity Cyano columns

Manufacturer: Thermo Scientific™ 22805012101

 View more versions of this product

Catalog No. 22805012101

  Check Availability
Add to Cart

Required Accessories : Thermo Scientific UNIGUARD direct connection or stand alone holder, sold separately.

    Specifications

    Particle Size 5μm
    Length (Metric) 10mm
    Diameter (Metric) Inner 2.1mm
    Stationary Phase Cyano

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit