Learn More
Thermo Scientific™ Immuno Standard Modules White and Black
For fluorescence/luminescence-based immunoassays and binding assays
Manufacturer:  Thermo Scientific™ 437877
Catalog No. 12-565-419
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Modules are available in 8-, 12- or 16-(2x8) well strips either in frames or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format gives highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format reduces back scatter light which may be encountered in epifluorescence
  • Use transparent or clear format in Time Resolved Fluorescence applications only
Choice of Surface:
  • PolySorp™ and Microlite™ 1+ for adsorption of hydrophobic molecules
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules
  • MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay
Specifications
Specifications
White, Black
PolySorp
8 x 12
Polystyrene
96
Standard