Thermo Scientific™ Jewett™ High-Performance Plasma Freezers
Designed to meet the highest standards for sensitive plasma freezing
$6936.00 - $15414.00
Specifications
Material High-impact, powder-coated paint
Metric Temperature Range -30°C
Metric Operating Temperature -30°C
Temperature Sensor Microprocessor
Door Type Solid
View More Specs
  • Products 7
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
JPL430A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
JPL430A
4.7 cu. ft. (133L); 115V 60Hz
Each for $6,936.00
Check Availability
Add to Cart
JPL1230A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
JPL1230A
11.5 cu. ft (326L); 115V 60Hz
Each for $9,420.00
Check Availability
Add to Cart
JPL2330A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
JPL2330A
23.3 cu. ft. (659L); 115V 60Hz
Each for $11,374.00
Check Availability
Add to Cart
JPL3030A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
JPL3030A
29.2 cu. ft. (826L); 115V 60Hz
Each for $12,699.00
Check Availability
Add to Cart
JPL3030D
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
JPL3030D
29.2 cu. ft. (826L); Holds 665 boxes; 1 door, 7 drawers; 208/240V, 60Hz
Each for $12,699.00
Check Availability
Add to Cart
JPL5030A
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
JPL5030A
51.1 cu. ft. (1447L); 115V 60Hz
Each for $15,414.00
Check Availability
Add to Cart
JPL5030D
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
JPL5030D
51.1 cu. ft. (1447L); 208/240V 60Hz
Each for $15,414.00
Check Availability
Add to Cart
Descriptions & Specifications
Related Products