Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Klenow Fragment of DNA Polymerase I

Removes overhanging 3« ends; fills in DNA fragments with overhanging 5«ends

Specifications

Concentration 5 U/μL
Product Type DNA Polymerase I
View More Specs

Includes: Separate 100mM MgCl2

Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
AB-0268
View Documents
Thermo Scientific™
AB0268
1000 Units 1000 Units Unknown catalog number. View substitute products »
Description

Description

  • Storage and Dilution Buffer: 50mM KH2PO2 (pH 7.0 at 25°C), 1mM DTT, 50% (v/v) glycerol
  • Reaction Buffer: 5X concentration. 250mM KH2PO2 (pH 7.0 at 25°C), 5mM DTT

Specifications

Specifications

5 U/μL
DNA Polymerase I
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit