Learn More
Thermo Scientific™ Cell Roller System Accessories for Environmental Chambers and Incubators
Allow production of monolayer cell cultures in standard roller culture vessels.
Manufacturer:  Thermo Scientific™ 190049
 View more versions of this product
Catalog No. 19-004-9
  Check Availability
Add to cart
Specifications
Description
Allow production of monolayer cell cultures in standard roller culture vessels with these Cell Roller System Accessories for Thermo Scientific™ Environmental Chambers and Incubators. Oxygenation and exposure of the cells to the media growth area are improved. Culture yields are increased by the uniform temperature control and cell roll system’s continuous, gentle rotation.
Description & Specifications
Specifications
Add-On Tier (5 positions)
Environmental Chambers and Incubators
24.4 x 29.8 x 7.1 in. (62 x 75.7 x 18cm)
Customer installed
Cell Roller System