Thermo Scientific™ Locator™ Cryogenic Rack & Box Systems
Inventory system that makes cell sample retrieval quick and easy
Manufacturer: Thermo Scientific™ HR509X2A
 View more versions of this product
Catalog No. 13-762-359
  Check Availability
Add to Cart
Required Accessories : Nalgene Cryogenic Storage Boxes (order separately). Each box has individually numbered storage areas for use with vials sized 2.0mL and smaller.
    Specifications
    Product Type Rack, Cryo
    For Use With Locator 4
  • Description & Specifications
Related Products
Product Content Feedback